The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001255982.1 | NP_001242911.1 | 3 - 4 | 562 | 89 |
NM_001255985.1 | NP_001242914.1 | 5 - 6 | 524 | 89 | |
NM_001255986.1 | NP_001242915.1 | 3 - 4 | 333 | 89 | |
NM_001255987.1 | NP_001242916.1 | 2 - 3 | 261 | 89 | |
NM_024027.4 | NP_076932.1 | 4 - 5 | 634 | 89 | |
NM_199235.2 | NP_954705.1 | 5 - 6 | 713 | 89 | |
NR_045659.1 | 4 - 5 | 634 | 89 | ||
XM_005263853.4 | XP_005263910.1 | 3 - 4 | 256 | 89 | |
GenBank mRNA | AB119525.1 | - | 4 - 5 | 520 | 89 |
AB119650.1 | - | 3 - 4 | 448 | 89 | |
AB119684.1 | - | 3 - 4 | 333 | 89 | |
AB119685.1 | - | 2 - 3 | 261 | 89 | |
AK303824.1 | - | 5 - 6 | 524 | 89 | |
AK313840.1 | - | 4 - 5 | 636 | 89 | |
AY358439.1 | - | 4 - 5 | 337 | 89 | |
BC000078.2 | - | 4 - 5 | 344 | 89 | |
BC009951.2 | - | 5 - 6 | 496 | 89 | |
BF316908.1 | - | 4 - 5 | 417 | 89 | |
BM547424.1 | - | 3 - 4 | 312 | 89 |