Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSSC1
Entrez Gene ID
7260
Gene Name
tumor suppressing subtransferable candidate 1
Gene Aliases
-
Chromosome Location
Chr.2: 3188968 - 3377882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003310.2 NP_003301.1 4 - 5 611 63
XM_006711893.2 XP_006711956.1 2 - 3 516 63
XM_011510383.2 XP_011508685.1 5 - 6 747 63
XM_011510384.2 XP_011508686.1 5 - 6 640 63
XM_011510385.2 XP_011508687.1 4 - 5 640 63
XM_017004818.1 XP_016860307.1 4 - 5 712 63
GenBank mRNA AF019952.1 - 4 - 5 569 63
AK026522.1 - 4 - 5 552 63
AK126288.1 - 7 - 8 1600 63
AK222652.1 - 4 - 5 547 63
BC002485.2 - 4 - 5 519 63

View Full Product Details