The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305102.1 | NP_001292031.1 | 13 - 14 | 1601 | 145 |
NM_007271.3 | NP_009202.1 | 13 - 14 | 1604 | 145 | |
XM_006714988.3 | XP_006715051.1 | 13 - 14 | 1984 | 145 | |
XM_006714989.3 | XP_006715052.1 | 13 - 14 | 1981 | 145 | |
GenBank mRNA | AK312559.1 | - | 13 - 14 | 1512 | 145 |
BC012085.1 | - | 13 - 14 | 1558 | 145 | |
BC095413.1 | - | 13 - 14 | 1564 | 145 | |
Z35102.1 | - | 12 - 13 | 1867 | 145 |