The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305102.1 | NP_001292031.1 | 2 - 3 | 458 | 74 |
NM_007271.3 | NP_009202.1 | 2 - 3 | 461 | 74 | |
XM_006714988.3 | XP_006715051.1 | 2 - 3 | 841 | 74 | |
XM_006714989.3 | XP_006715052.1 | 2 - 3 | 838 | 74 | |
XM_017010226.1 | XP_016865715.1 | 2 - 3 | 421 | 74 | |
GenBank mRNA | AK312559.1 | - | 2 - 3 | 369 | 74 |
BC012085.1 | - | 2 - 3 | 415 | 74 | |
BC095413.1 | - | 2 - 3 | 421 | 74 | |
DA459946.1 | - | 2 - 3 | 458 | 74 | |
Z35102.1 | - | 1 - 2 | 724 | 74 |