The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305102.1 | NP_001292031.1 | 4 - 5 | 632 | 137 |
NM_007271.3 | NP_009202.1 | 4 - 5 | 635 | 137 | |
XM_006714988.3 | XP_006715051.1 | 4 - 5 | 1015 | 137 | |
XM_006714989.3 | XP_006715052.1 | 4 - 5 | 1012 | 137 | |
XM_017010226.1 | XP_016865715.1 | 4 - 5 | 595 | 137 | |
GenBank mRNA | AF034188.1 | - | 3 - 4 | 298 | 137 |
AK312559.1 | - | 4 - 5 | 543 | 137 | |
BC012085.1 | - | 4 - 5 | 589 | 137 | |
BC095413.1 | - | 4 - 5 | 595 | 137 | |
Z35102.1 | - | 3 - 4 | 898 | 137 |