The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139033.2 | NP_620602.2 | 1 - 2 | 383 | 65 |
XM_006721558.3 | XP_006721621.1 | 1 - 2 | 383 | 65 | |
GenBank mRNA | AK295376.1 | - | 1 - 2 | 77 | 65 |
DB133227.1 | - | 1 - 2 | 384 | 65 | |
U29725.1 | - | 1 - 2 | 218 | 65 |