Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FURIN
Entrez Gene ID
5045
Gene Name
furin, paired basic amino acid cleaving enzyme
Gene Aliases
FUR, PACE, PCSK3, SPC1
Chromosome Location
Chr.15: 90868592 - 90883458 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289823.1 NP_001276752.1 11 - 12 1490 70
NM_001289824.1 NP_001276753.1 11 - 12 1644 70
NM_002569.3 NP_002560.1 11 - 12 1543 70
GenBank mRNA AL110107.1 - 3 - 4 385 70
BC012181.1 - 10 - 11 1313 70
X17094.1 - 11 - 12 1480 70

View Full Product Details