The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289823.1 | NP_001276752.1 | 6 - 7 | 800 | 91 |
NM_001289824.1 | NP_001276753.1 | 6 - 7 | 954 | 91 | |
NM_002569.3 | NP_002560.1 | 6 - 7 | 853 | 91 | |
GenBank mRNA | BC012181.1 | - | 5 - 6 | 623 | 91 |
X17094.1 | - | 6 - 7 | 790 | 91 |