The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005108.3 | NP_005099.2 | 16 - 17 | 1453 | 113 |
XM_011534325.2 | XP_011532627.1 | 16 - 17 | 1434 | 113 | |
XM_011534327.1 | XP_011532629.1 | 16 - 17 | 1434 | 113 | |
XM_011534328.2 | XP_011532630.1 | 16 - 17 | 1434 | 113 | |
XM_011534329.1 | XP_011532631.1 | 16 - 17 | 1434 | 113 | |
XM_011534330.2 | XP_011532632.1 | 16 - 17 | 1434 | 113 | |
XM_017007595.1 | XP_016863084.1 | 14 - 15 | 1202 | 113 | |
XM_017007596.1 | XP_016863085.1 | 16 - 17 | 1479 | 113 | |
XM_017007597.1 | XP_016863086.1 | 8 - 9 | 768 | 113 | |
XM_017007598.1 | XP_016863087.1 | 16 - 17 | 1434 | 113 | |
XM_017007599.1 | XP_016863088.1 | 15 - 16 | 1337 | 113 | |
XM_017007600.1 | XP_016863089.1 | 13 - 14 | 1247 | 113 | |
GenBank mRNA | AB015046.1 | - | 16 - 17 | 1473 | 113 |
AK001205.1 | - | 9 - 10 | 724 | 113 | |
AK025728.1 | - | 16 - 17 | 1405 | 113 | |
AK293325.1 | - | 14 - 15 | 1207 | 113 | |
AK314386.1 | - | 16 - 17 | 1401 | 113 | |
BC137076.1 | - | 16 - 17 | 1385 | 113 | |
BC137080.1 | - | 16 - 17 | 1385 | 113 |