The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005108.3 | NP_005099.2 | 8 - 9 | 755 | 79 |
XM_011534325.2 | XP_011532627.1 | 8 - 9 | 736 | 79 | |
XM_011534327.1 | XP_011532629.1 | 8 - 9 | 736 | 79 | |
XM_011534328.2 | XP_011532630.1 | 8 - 9 | 736 | 79 | |
XM_011534329.1 | XP_011532631.1 | 8 - 9 | 736 | 79 | |
XM_011534330.2 | XP_011532632.1 | 8 - 9 | 736 | 79 | |
XM_017007595.1 | XP_016863084.1 | 6 - 7 | 504 | 79 | |
XM_017007596.1 | XP_016863085.1 | 8 - 9 | 781 | 79 | |
XM_017007598.1 | XP_016863087.1 | 8 - 9 | 736 | 79 | |
XM_017007599.1 | XP_016863088.1 | 8 - 9 | 736 | 79 | |
XM_017007600.1 | XP_016863089.1 | 6 - 7 | 590 | 79 | |
GenBank mRNA | AB015046.1 | - | 8 - 9 | 775 | 79 |
AK025728.1 | - | 8 - 9 | 707 | 79 | |
AK293325.1 | - | 6 - 7 | 509 | 79 | |
AK314386.1 | - | 8 - 9 | 703 | 79 | |
BC137076.1 | - | 8 - 9 | 687 | 79 | |
BC137080.1 | - | 8 - 9 | 687 | 79 |