The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005108.3 | NP_005099.2 | 18 - 19 | 1641 | 89 |
GenBank mRNA | AK001205.1 | - | 11 - 12 | 912 | 89 |
AK022601.1 | - | 1 - 2 | 98 | 89 | |
AK025728.1 | - | 18 - 19 | 1593 | 89 | |
AK293325.1 | - | 16 - 17 | 1395 | 89 | |
AK314386.1 | - | 18 - 19 | 1589 | 89 | |
BC137076.1 | - | 18 - 19 | 1573 | 89 | |
BC137080.1 | - | 18 - 19 | 1573 | 89 |