The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005108.3 | NP_005099.2 | 2 - 3 | 66 | |
XM_011534325.2 | XP_011532627.1 | 2 - 3 | 234 | 66 | |
XM_011534327.1 | XP_011532629.1 | 2 - 3 | 234 | 66 | |
XM_011534328.2 | XP_011532630.1 | 2 - 3 | 234 | 66 | |
XM_011534329.1 | XP_011532631.1 | 2 - 3 | 234 | 66 | |
XM_011534330.2 | XP_011532632.1 | 2 - 3 | 234 | 66 | |
XM_017007596.1 | XP_016863085.1 | 2 - 3 | 279 | 66 | |
XM_017007598.1 | XP_016863087.1 | 2 - 3 | 234 | 66 | |
XM_017007599.1 | XP_016863088.1 | 2 - 3 | 234 | 66 | |
GenBank mRNA | AK025728.1 | - | 2 - 3 | 66 | |
AK314386.1 | - | 2 - 3 | 66 | ||
BC030231.2 | - | 2 - 3 | 66 | ||
BC137076.1 | - | 2 - 3 | 66 | ||
BC137080.1 | - | 2 - 3 | 66 | ||
BI090475.1 | - | 2 - 3 | 66 |