Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RING1
Entrez Gene ID
6015
Gene Name
ring finger protein 1
Gene Aliases
RING1A, RNF1
Chromosome Location
Chr.6: 33208509 - 33212722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002931.3 NP_002922.2 4 - 5 647 76
GenBank mRNA AK289355.1 - 4 - 5 656 76
AK301010.1 - 4 - 5 656 76
AK301378.1 - 4 - 5 590 76
BC002922.2 - 4 - 5 642 76
BC051866.2 - 4 - 5 640 76
BT007352.1 - 2 - 3 366 76
Z14000.1 - 2 - 3 441 76

View Full Product Details