Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D22B
Entrez Gene ID
55633
Gene Name
TBC1 domain family member 22B
Gene Aliases
C6orf197
Chromosome Location
Chr.6: 37257704 - 37332970 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017772.3 NP_060242.2 12 - 13 1601 80
NR_130108.1 12 - 13 1662 80
GenBank mRNA AB449909.1 - 12 - 13 1387 80
AK000344.1 - 5 - 6 508 80
AK096340.1 - 12 - 13 1530 80
AK292893.1 - 12 - 13 1496 80
AY313781.1 - 12 - 13 1533 80
AY927466.1 - 5 - 6 682 80
BC000291.2 - 3 - 4 255 80
BC000743.2 - 3 - 4 255 80
BC001927.1 - 3 - 4 255 80
BC002720.2 - 5 - 6 461 80
BC047658.1 - 12 - 13 1594 80
BC063523.1 - 7 - 8 611 80
BC109026.1 - 12 - 13 1531 80
BC109027.1 - 12 - 13 1531 80
BX648317.1 - 12 - 13 1532 80
CR749710.1 - 6 - 7 687 80

View Full Product Details