The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012343.3 | NP_036475.3 | 18 - 19 | 3045 | 72 |
NM_182977.2 | NP_892022.2 | 18 - 19 | 3011 | 72 | |
XM_005248274.4 | XP_005248331.1 | 18 - 19 | 3023 | 72 | |
XM_005248275.4 | XP_005248332.1 | 13 - 14 | 2273 | 72 | |
XM_006714461.3 | XP_006714524.1 | 17 - 18 | 2637 | 72 | |
XM_011514001.2 | XP_011512303.1 | 18 - 19 | 3042 | 72 | |
XM_011514002.2 | XP_011512304.1 | 17 - 18 | 2732 | 72 | |
XM_017009293.1 | XP_016864782.1 | 18 - 19 | 2913 | 72 | |
GenBank mRNA | AK297558.1 | - | 11 - 12 | 1658 | 72 |
AK299622.1 | - | 10 - 11 | 1475 | 72 | |
AL831822.2 | - | 18 - 19 | 3012 | 72 | |
BC032370.1 | - | 2 - 3 | 240 | 72 | |
BC110543.1 | - | 18 - 19 | 2861 | 72 | |
BC110544.1 | - | 17 - 18 | 2858 | 72 | |
Z50101.1 | - | 17 - 18 | 2833 | 72 |