Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C2CD2
Entrez Gene ID
25966
Gene Name
C2 calcium dependent domain containing 2
Gene Aliases
C21orf25, C21orf258, TMEM24L
Chromosome Location
Chr.21: 41885110 - 41953957 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011529523.2 XP_011527825.1 1 - 2 150 92

View Full Product Details