Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMA5
Entrez Gene ID
3911
Gene Name
laminin subunit alpha 5
Gene Aliases
-
Chromosome Location
Chr.20: 62309060 - 62367318 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005560.4 NP_005551.3 11 - 12 1549 142
XM_006723796.3 XP_006723859.1 11 - 12 1552 142
XM_006723798.3 XP_006723861.1 11 - 12 1555 142
XM_011528818.2 XP_011527120.1 11 - 12 1552 142
XM_011528819.2 XP_011527121.1 11 - 12 1552 142
GenBank mRNA AB067494.1 - 8 - 9 893 142
AF443072.1 - 11 - 12 1549 142

View Full Product Details