Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMA5
Entrez Gene ID
3911
Gene Name
laminin subunit alpha 5
Gene Aliases
-
Chromosome Location
Chr.20: 62309060 - 62367318 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005560.4 NP_005551.3 60 - 61 8269 75
XM_006723796.3 XP_006723859.1 60 - 61 8272 75
XM_011528818.2 XP_011527120.1 59 - 60 8134 75
XM_011528819.2 XP_011527121.1 58 - 59 7999 75
GenBank mRNA AB011105.1 - 15 - 16 2054 75
AF443072.1 - 60 - 61 8269 75

View Full Product Details