Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMA5
Entrez Gene ID
3911
Gene Name
laminin subunit alpha 5
Gene Aliases
-
Chromosome Location
Chr.20: 62309060 - 62367318 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005560.4 NP_005551.3 63 - 64 8724 106
XM_006723796.3 XP_006723859.1 63 - 64 8727 106
XM_011528818.2 XP_011527120.1 62 - 63 8589 106
XM_011528819.2 XP_011527121.1 61 - 62 8454 106
GenBank mRNA AB011105.1 - 18 - 19 2509 106
AF443072.1 - 63 - 64 8724 106
Z95636.1 - 3 - 4 431 106

View Full Product Details