The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015268.3 | NP_056083.3 | 3 - 4 | 209 | 92 |
XM_005247245.4 | XP_005247302.1 | 3 - 4 | 391 | 92 | |
GenBank mRNA | AB014578.2 | - | 3 - 4 | 258 | 92 |
AY779857.1 | - | 3 - 4 | 209 | 92 | |
BC043583.1 | - | 3 - 4 | 209 | 92 | |
BC151246.1 | - | 4 - 5 | 279 | 92 |