The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015268.3 | NP_056083.3 | 43 - 44 | 5174 | 132 |
XM_005247245.4 | XP_005247302.1 | 44 - 45 | 5371 | 132 | |
XM_017006036.1 | XP_016861525.1 | 30 - 31 | 3631 | 132 | |
GenBank mRNA | AB014578.2 | - | 43 - 44 | 5223 | 132 |
AK023168.1 | - | 12 - 13 | 1602 | 132 | |
AK125330.1 | - | 20 - 21 | 2558 | 132 | |
AK127112.1 | - | 27 - 28 | 3262 | 132 | |
AY779857.1 | - | 43 - 44 | 5174 | 132 | |
BC151246.1 | - | 44 - 45 | 5244 | 132 |