The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146102.1 | NP_001139574.1 | 5 - 6 | 1352 | 114 |
NM_001146103.1 | NP_001139575.1 | 5 - 6 | 1429 | 114 | |
NM_001146104.1 | NP_001139576.1 | 5 - 6 | 1428 | 114 | |
NM_001146105.1 | NP_001139577.1 | 5 - 6 | 1247 | 114 | |
NM_001146106.1 | NP_001139578.1 | 5 - 6 | 1428 | 114 | |
NM_031458.2 | NP_113646.2 | 5 - 6 | 1534 | 114 | |
XM_005247820.2 | XP_005247877.1 | 5 - 6 | 1458 | 114 | |
XM_011513218.1 | XP_011511520.1 | 5 - 6 | 1324 | 114 | |
XM_017007304.1 | XP_016862793.1 | 5 - 6 | 1342 | 114 | |
XM_017007305.1 | XP_016862794.1 | 5 - 6 | 1237 | 114 | |
XM_017007306.1 | XP_016862795.1 | 5 - 6 | 1318 | 114 | |
GenBank mRNA | AB209742.1 | - | 1 - 2 | 51 | 114 |
AF307338.1 | - | 5 - 6 | 1435 | 114 | |
AF307339.1 | - | 5 - 6 | 1330 | 114 | |
AK292959.1 | - | 5 - 6 | 1207 | 114 | |
AK299107.1 | - | 5 - 6 | 1321 | 114 | |
AK313494.1 | - | 5 - 6 | 1321 | 114 | |
BC017463.1 | - | 5 - 6 | 1396 | 114 | |
BC039580.1 | - | 4 - 5 | 1205 | 114 |