The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114121.2 | NP_001107593.1 | 6 - 7 | 1510 | 79 |
NM_001114122.2 | NP_001107594.1 | 6 - 7 | 1510 | 79 | |
NM_001244846.1 | NP_001231775.1 | 6 - 7 | 1510 | 79 | |
NM_001274.5 | NP_001265.2 | 6 - 7 | 853 | 79 | |
NR_045204.1 | 5 - 6 | 1286 | 79 | ||
NR_045205.1 | 6 - 7 | 1047 | 79 | ||
XM_011542560.2 | XP_011540862.1 | 5 - 6 | 1512 | 79 | |
XM_011542561.2 | XP_011540863.1 | 5 - 6 | 1512 | 79 | |
XM_011542562.2 | XP_011540864.1 | 5 - 6 | 762 | 79 | |
XM_011542563.2 | XP_011540865.1 | 5 - 6 | 827 | 79 | |
XM_017017146.1 | XP_016872635.1 | 6 - 7 | 697 | 79 | |
GenBank mRNA | AB451222.1 | - | 5 - 6 | 613 | 79 |
AB451345.1 | - | 5 - 6 | 613 | 79 | |
AF016582.1 | - | 5 - 6 | 647 | 79 | |
AF032874.1 | - | 5 - 6 | 613 | 79 | |
AK292549.1 | - | 6 - 7 | 1047 | 79 | |
AK293143.1 | - | 6 - 7 | 675 | 79 | |
AK299783.1 | - | 5 - 6 | 447 | 79 | |
AK300082.1 | - | 5 - 6 | 696 | 79 | |
BC004202.2 | - | 6 - 7 | 672 | 79 | |
BC017575.1 | - | 6 - 7 | 771 | 79 | |
JF289264.1 | - | 5 - 6 | 488 | 79 |