The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002379.3 | NP_002370.1 | 4 - 5 | 829 | 78 |
GenBank mRNA | AK075519.1 | - | 4 - 5 | 832 | 78 |
AK312949.1 | - | 4 - 5 | 829 | 78 | |
BC131710.1 | - | 4 - 5 | 747 | 78 | |
M55683.1 | - | 2 - 3 | 327 | 78 |