Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SZT2
Entrez Gene ID
23334
Gene Name
seizure threshold 2 homolog (mouse)
Gene Aliases
C1orf84, EIEE18, KIAA0467, SZT2A, SZT2B
Chromosome Location
Chr.1: 43389884 - 43454247 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015284.3 NP_056099.3 70 - 71 10067 75
XM_005270686.3 XP_005270743.1 72 - 73 10299 75
XM_006710501.3 XP_006710564.1 71 - 72 10186 75
XM_011541106.2 XP_011539408.1 71 - 72 10337 75
XM_011541107.2 XP_011539409.1 68 - 69 9834 75
XM_017000819.1 XP_016856308.1 72 - 73 10146 75
XM_017000820.1 XP_016856309.1 71 - 72 10128 75
XM_017000821.1 XP_016856310.1 68 - 69 9820 75
GenBank mRNA AB007936.2 - 56 - 57 7919 75
AK001249.2 - 5 - 6 878 75
AK027078.1 - 11 - 12 1612 75
AK126972.1 - 9 - 10 1969 75
AL117402.1 - 5 - 6 861 75
BC041069.1 - 12 - 13 1748 75
BC052802.1 - 13 - 14 1762 75
BC082968.1 - 13 - 14 1767 75
BC151232.1 - 56 - 57 7919 75

View Full Product Details