The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014046.3 | NP_054765.1 | 2 - 3 | 343 | 135 |
GenBank mRNA | AF100761.1 | - | 2 - 3 | 198 | 135 |
AF151017.1 | - | 2 - 3 | 224 | 135 | |
AK294086.1 | - | 2 - 3 | 205 | 135 | |
AK311890.1 | - | 2 - 3 | 194 | 135 | |
BC005373.1 | - | 2 - 3 | 223 | 135 | |
BG773408.1 | - | 2 - 3 | 334 | 135 | |
BP369345.1 | - | 2 - 3 | 343 | 135 |