The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199165.2 | NP_001186094.1 | 21 - 22 | 2539 | 79 |
NM_001302891.1 | NP_001289820.1 | 19 - 20 | 2489 | 79 | |
XM_005257119.4 | XP_005257176.1 | 21 - 22 | 2633 | 79 | |
XM_005257125.3 | XP_005257182.1 | 13 - 14 | 1623 | 79 | |
XM_005257126.4 | XP_005257183.1 | 10 - 11 | 1288 | 79 | |
XM_006721740.2 | XP_006721803.1 | 21 - 22 | 2505 | 79 | |
XM_011524461.2 | XP_011522763.1 | 21 - 22 | 2646 | 79 | |
XM_011524462.2 | XP_011522764.1 | 21 - 22 | 2635 | 79 | |
XM_011524463.2 | XP_011522765.1 | 21 - 22 | 2635 | 79 | |
XM_011524465.2 | XP_011522767.1 | 18 - 19 | 1962 | 79 | |
XM_011524466.2 | XP_011522768.1 | 11 - 12 | 1441 | 79 | |
XM_011524467.2 | XP_011522769.1 | 10 - 11 | 1283 | 79 | |
XM_017024302.1 | XP_016879791.1 | 19 - 20 | 2376 | 79 | |
GenBank mRNA | AF458591.1 | - | 11 - 12 | 1446 | 79 |
AK299075.1 | - | 21 - 22 | 2490 | 79 | |
BC131805.1 | - | 19 - 20 | 2317 | 79 | |
BX097629.1 | - | 1 - 2 | 86 | 79 | |
CX872964.1 | - | 5 - 6 | 732 | 79 |