Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP112
Entrez Gene ID
201134
Gene Name
centrosomal protein 112
Gene Aliases
CCDC46, MACOCO
Chromosome Location
Chr.17: 65635540 - 66192131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037325.2 NP_001032402.1 6 - 7 876 99
NM_001199165.2 NP_001186094.1 26 - 27 3007 99
NM_001302891.1 NP_001289820.1 24 - 25 2957 99
NR_126542.1 5 - 6 706 99
XM_005257119.4 XP_005257176.1 26 - 27 3101 99
XM_005257125.3 XP_005257182.1 18 - 19 2091 99
XM_005257126.4 XP_005257183.1 15 - 16 1756 99
XM_006721740.2 XP_006721803.1 26 - 27 2973 99
XM_011524461.2 XP_011522763.1 26 - 27 3114 99
XM_011524464.2 XP_011522766.1 24 - 25 2890 99
XM_011524465.2 XP_011522767.1 23 - 24 2430 99
XM_011524466.2 XP_011522768.1 16 - 17 1909 99
XM_011524467.2 XP_011522769.1 15 - 16 1751 99
XM_017024302.1 XP_016879791.1 24 - 25 2844 99
GenBank mRNA AA781351.1 - 1 - 2 99
AF458591.1 - 16 - 17 1914 99
AK098070.1 - 16 - 17 1826 99
AK299075.1 - 26 - 27 2958 99
BC029524.1 - 6 - 7 876 99
BC131805.1 - 24 - 25 2785 99
BG203878.1 - - 173 99
BG531260.1 - 3 - 4 194 99
BX097629.1 - 6 - 7 554 99

View Full Product Details