The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014550.3 | NP_055365.2 | 15 - 16 | 2374 | 91 |
GenBank mRNA | AK074906.1 | - | 12 - 13 | 1604 | 91 |
AK296176.1 | - | 3 - 4 | 644 | 91 | |
AY028896.1 | - | 15 - 16 | 2411 | 91 | |
AY032928.1 | - | 15 - 16 | 2374 | 91 | |
BC032058.1 | - | 13 - 14 | 1999 | 91 | |
BC113659.1 | - | 15 - 16 | 2392 | 91 | |
BC117403.1 | - | 15 - 16 | 2386 | 91 |