The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317009.1 | NP_001303938.1 | 5 - 6 | 2000 | 90 |
NM_053277.2 | NP_444507.1 | 4 - 5 | 1946 | 90 | |
XM_017028406.1 | XP_016883895.1 | 4 - 5 | 2156 | 90 | |
GenBank mRNA | AF426169.1 | - | 1 - 2 | 77 | 90 |
AF448438.1 | - | 4 - 5 | 1720 | 90 | |
AF448439.1 | - | 5 - 6 | 1774 | 90 | |
AK092733.1 | - | 3 - 4 | 261 | 90 | |
AK289663.1 | - | 4 - 5 | 1946 | 90 | |
BC040196.2 | - | 4 - 5 | 886 | 90 |