The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003642.3 | NP_003633.1 | 2 - 3 | 187 | 116 |
XM_006712808.3 | XP_006712871.1 | 3 - 4 | 383 | 116 | |
XM_011512027.2 | XP_011510329.1 | 2 - 3 | 939 | 116 | |
XM_017005136.1 | XP_016860625.1 | 3 - 4 | 1160 | 116 | |
XM_017005137.1 | XP_016860626.1 | 3 - 4 | 1092 | 116 | |
GenBank mRNA | AF030424.1 | - | 2 - 3 | 147 | 116 |
AK313699.1 | - | 2 - 3 | 177 | 117 | |
DB050336.1 | - | 3 - 4 | 388 | 116 |