The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003642.3 | NP_003633.1 | 9 - 10 | 1052 | 94 |
NR_027862.1 | 8 - 9 | 976 | 94 | ||
GenBank mRNA | AF030424.1 | - | 9 - 10 | 1012 | 94 |
AK127840.1 | - | 3 - 4 | 2001 | 94 | |
AK309001.1 | - | 8 - 9 | 894 | 94 | |
AK313699.1 | - | 9 - 10 | 1043 | 94 | |
BC018682.1 | - | 5 - 6 | 528 | 94 | |
BC063003.1 | - | 8 - 9 | 953 | 94 |