The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002931.3 | NP_002922.2 | 1 - 2 | 134 | 152 |
GenBank mRNA | AK289355.1 | - | 1 - 2 | 143 | 152 |
AK297552.1 | - | 1 - 2 | 133 | 152 | |
AK301010.1 | - | 1 - 2 | 143 | 152 | |
BC002922.2 | - | 1 - 2 | 129 | 152 | |
BC051866.2 | - | 1 - 2 | 127 | 152 | |
BI259576.1 | - | 1 - 2 | 143 | 152 |