Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TULP1
Entrez Gene ID
7287
Gene Name
tubby like protein 1
Gene Aliases
LCA15, RP14, TUBL1
Chromosome Location
Chr.6: 35497874 - 35512902 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289395.1 NP_001276324.1 11 - 12 1110 124
NM_003322.4 NP_003313.3 12 - 13 1269 124
GenBank mRNA AB593124.1 - 12 - 13 1255 124
BC032714.1 - 11 - 12 1095 124
BC065261.1 - 11 - 12 1078 124
U82468.1 - 12 - 13 1261 124

View Full Product Details