The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003322.4 | NP_003313.3 | 3 - 4 | 237 | 121 |
GenBank mRNA | AB593123.1 | - | 3 - 4 | 121 | |
AB593124.1 | - | 3 - 4 | 121 | ||
U82468.1 | - | 3 - 4 | 121 |