Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TULP1
Entrez Gene ID
7287
Gene Name
tubby like protein 1
Gene Aliases
LCA15, RP14, TUBL1
Chromosome Location
Chr.6: 35497874 - 35512902 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289395.1 NP_001276324.1 9 - 10 888 85
NM_003322.4 NP_003313.3 10 - 11 1047 85
GenBank mRNA AB593123.1 - 9 - 10 1044 85
AB593124.1 - 10 - 11 1033 85
BC032714.1 - 9 - 10 873 85
BC065261.1 - 9 - 10 856 85
U82468.1 - 10 - 11 1039 85

View Full Product Details