The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000868.3 | NP_000859.1 | 1 - 2 | 583 | 59 |
NM_001256760.2 | NP_001243689.1 | 1 - 2 | 583 | 59 | |
NM_001256761.2 | NP_001243690.1 | 1 - 2 | 583 | 59 | |
GenBank mRNA | AK295753.1 | - | 1 - 2 | 575 | 59 |
AK308766.1 | - | 1 - 2 | 162 | 59 | |
BC028688.1 | - | 2 - 3 | 536 | 59 | |
M81778.1 | - | 1 - 2 | 551 | 59 | |
U49516.1 | - | 1 - 2 | 583 | 59 |