Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRM6
Entrez Gene ID
2916
Gene Name
glutamate metabotropic receptor 6
Gene Aliases
CSNB1B, GPRC1F, MGLUR6, mGlu6
Chromosome Location
Chr.5: 178978327 - 178995123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000843.3 NP_000834.2 9 - 10 2610 113

View Full Product Details