The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003386.2 | NP_003377.2 | 25 - 26 | 5026 | 95 |
NM_173059.2 | NP_775082.2 | 25 - 26 | 5026 | 95 | |
NR_111917.1 | 25 - 26 | 5026 | 95 | ||
NR_111918.1 | 25 - 26 | 5026 | 95 | ||
NR_111919.1 | 25 - 26 | 5026 | 95 | ||
XM_011516555.2 | XP_011514857.1 | 25 - 26 | 4759 | 95 | |
XM_017012581.1 | XP_016868070.1 | 12 - 13 | 2515 | 95 | |
GenBank mRNA | AF332975.1 | - | 25 - 26 | 5026 | 95 |
AF332976.1 | - | 25 - 26 | 5026 | 95 | |
AF332977.1 | - | 25 - 26 | 5026 | 95 | |
AF332978.1 | - | 25 - 26 | 5026 | 95 | |
AF332979.1 | - | 25 - 26 | 5026 | 95 | |
AF332980.1 | - | 25 - 26 | 5026 | 95 | |
AK302596.1 | - | 4 - 5 | 654 | 95 |