The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004239.4 | NP_004230.2 | 1 - 2 | 514 | 104 |
XM_017021787.1 | XP_016877276.1 | 1 - 2 | 494 | 104 | |
XM_017021788.1 | XP_016877277.1 | 1 - 2 | 514 | 104 | |
GenBank mRNA | AF007217.1 | - | 1 - 2 | 491 | 104 |
AK307489.1 | - | 1 - 2 | 514 | 104 | |
AL599917.1 | - | 1 - 2 | 506 | 104 | |
BC002656.2 | - | 1 - 2 | 480 | 104 | |
BC069008.1 | - | 1 - 2 | 488 | 104 | |
BC146845.1 | - | 1 - 2 | 396 | 104 | |
Y12490.1 | - | 1 - 2 | 496 | 104 |