Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TET2
Entrez Gene ID
54790
Gene Name
tet methylcytosine dioxygenase 2
Gene Aliases
KIAA1546, MDS
Chromosome Location
Chr.4: 105145875 - 105279803 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127208.2 NP_001120680.1 4 - 5 3987 125
XM_005263082.2 XP_005263139.1 3 - 4 3863 125
GenBank mRNA DR000714.1 - 2 - 3 203 125
FM992369.1 - 4 - 5 4360 125

View Full Product Details