Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARD3
Entrez Gene ID
56288
Gene Name
par-3 family cell polarity regulator
Gene Aliases
ASIP, Baz, PAR3, PAR3alpha, PARD-3, PARD3A, PPP1R118, SE2-5L16, SE2-5LT1, SE2-5T2
Chromosome Location
Chr.10: 34109560 - 34815325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184785.1 NP_001171714.1 2 - 3 553 104
NM_001184786.1 NP_001171715.1 2 - 3 553 104
NM_001184787.1 NP_001171716.1 2 - 3 553 104
NM_001184788.1 NP_001171717.1 2 - 3 553 104
NM_001184789.1 NP_001171718.1 2 - 3 553 104
NM_001184790.1 NP_001171719.1 2 - 3 553 104
NM_001184791.1 NP_001171720.1 2 - 3 553 104
NM_001184792.1 NP_001171721.1 2 - 3 553 104
NM_001184793.1 NP_001171722.1 2 - 3 553 104
NM_001184794.1 NP_001171723.1 2 - 3 553 104
NM_019619.3 NP_062565.2 2 - 3 553 104
XM_005252528.4 XP_005252585.1 2 - 3 544 104
XM_005252530.4 XP_005252587.1 2 - 3 544 104
XM_005252531.4 XP_005252588.1 2 - 3 544 104
XM_005252532.4 XP_005252589.1 2 - 3 330 104
XM_005252534.4 XP_005252591.1 2 - 3 544 104
XM_005252535.4 XP_005252592.1 2 - 3 544 104
XM_005252536.4 XP_005252593.1 2 - 3 544 104
XM_011519569.2 XP_011517871.1 2 - 3 544 104
XM_011519570.2 XP_011517872.1 2 - 3 544 104
XM_011519571.2 XP_011517873.1 2 - 3 544 104
XM_011519572.2 XP_011517874.1 2 - 3 544 104
XM_011519573.2 XP_011517875.1 2 - 3 544 104
XM_011519576.2 XP_011517878.1 2 - 3 331 104
XM_011519577.2 XP_011517879.1 2 - 3 544 104
XM_011519578.2 XP_011517880.1 2 - 3 548 104
XM_011519581.2 XP_011517883.1 2 - 3 544 104
XM_011519583.2 XP_011517885.1 2 - 3 544 104
XM_011519584.2 XP_011517886.1 2 - 3 544 104
XM_017016430.1 XP_016871919.1 2 - 3 544 104
GenBank mRNA AB073671.1 - 2 - 3 223 104
AF196185.1 - 2 - 3 510 104
AF196186.1 - 2 - 3 510 104
AF252293.1 - 2 - 3 223 104
AF332592.1 - 2 - 3 223 104
AF332593.1 - 2 - 3 223 104
AF454058.1 - 2 - 3 567 104
AF467002.1 - 2 - 3 223 104
AF467003.1 - 2 - 3 223 104
AF467004.1 - 2 - 3 223 104
AF467005.1 - 2 - 3 223 104
AF467006.1 - 2 - 3 223 104
BC071566.1 - 2 - 3 500 104

View Full Product Details