Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARD3
Entrez Gene ID
56288
Gene Name
par-3 family cell polarity regulator
Gene Aliases
ASIP, Baz, PAR3, PAR3alpha, PARD-3, PARD3A, PPP1R118, SE2-5L16, SE2-5LT1, SE2-5T2
Chromosome Location
Chr.10: 34109560 - 34815325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184785.1 NP_001171714.1 9 - 10 1732 145
NM_001184786.1 NP_001171715.1 9 - 10 1732 145
NM_001184787.1 NP_001171716.1 9 - 10 1732 145
NM_001184788.1 NP_001171717.1 9 - 10 1732 145
NM_001184789.1 NP_001171718.1 9 - 10 1732 145
NM_001184790.1 NP_001171719.1 8 - 9 1600 145
NM_001184791.1 NP_001171720.1 8 - 9 1600 145
NM_001184792.1 NP_001171721.1 9 - 10 1732 145
NM_001184793.1 NP_001171722.1 9 - 10 1732 145
NM_001184794.1 NP_001171723.1 9 - 10 1732 145
NM_019619.3 NP_062565.2 9 - 10 1732 145
XM_005252528.4 XP_005252585.1 9 - 10 1723 145
XM_005252530.4 XP_005252587.1 9 - 10 1723 145
XM_005252531.4 XP_005252588.1 9 - 10 1723 145
XM_005252532.4 XP_005252589.1 8 - 9 1377 145
XM_005252534.4 XP_005252591.1 9 - 10 1723 145
XM_005252535.4 XP_005252592.1 9 - 10 1723 145
XM_005252536.4 XP_005252593.1 9 - 10 1723 145
XM_011519569.2 XP_011517871.1 9 - 10 1723 145
XM_011519570.2 XP_011517872.1 9 - 10 1723 145
XM_011519571.2 XP_011517873.1 9 - 10 1723 145
XM_011519572.2 XP_011517874.1 9 - 10 1723 145
XM_011519573.2 XP_011517875.1 9 - 10 1723 145
XM_011519575.2 XP_011517877.1 8 - 9 1407 145
XM_011519576.2 XP_011517878.1 8 - 9 1378 145
XM_011519577.2 XP_011517879.1 9 - 10 1723 145
XM_011519578.2 XP_011517880.1 8 - 9 1595 145
XM_011519579.2 XP_011517881.1 7 - 8 1276 145
XM_011519581.2 XP_011517883.1 9 - 10 1723 145
XM_011519583.2 XP_011517885.1 9 - 10 1723 145
XM_011519584.2 XP_011517886.1 9 - 10 1723 145
XM_017016430.1 XP_016871919.1 9 - 10 1723 145
GenBank mRNA AB073671.1 - 8 - 9 1270 145
AF177228.1 - 2 - 3 468 145
AF196185.1 - 9 - 10 1689 145
AF196186.1 - 9 - 10 1689 145
AF252293.1 - 8 - 9 1270 145
AF332592.1 - 9 - 10 1402 145
AF332593.1 - 9 - 10 1402 145
AF454057.1 - 7 - 8 1109 145
AF454058.1 - 8 - 9 1614 145
AF454059.1 - 5 - 6 735 145
AF467002.1 - 9 - 10 1402 145
AF467003.1 - 9 - 10 1402 145
AF467004.1 - 9 - 10 1402 145
AF467005.1 - 8 - 9 1270 145
AF467006.1 - 9 - 10 1402 145
AK025892.1 - 1 - 2 218 145
AK027735.1 - 7 - 8 1029 145
BC071566.1 - 9 - 10 1679 145

View Full Product Details