The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001249.3 | NP_001240.1 | 1 - 2 | 71 | 60 |
NM_001321984.1 | NP_001308913.1 | 1 - 2 | 71 | 60 | |
NM_001321985.1 | NP_001308914.1 | 1 - 2 | 71 | 60 | |
NM_001321986.1 | NP_001308915.1 | 1 - 2 | 71 | 60 | |
NM_001321987.1 | NP_001308916.1 | 1 - 2 | 71 | 60 | |
XM_005268224.3 | XP_005268281.1 | 1 - 2 | 57 | 60 | |
XM_006720325.3 | XP_006720388.1 | 1 - 2 | 74 | 60 | |
XM_006720326.2 | XP_006720389.1 | 1 - 2 | 74 | 60 | |
XM_017021813.1 | XP_016877302.1 | 1 - 2 | 66 | 60 | |
XM_017021815.1 | XP_016877304.1 | 1 - 2 | 66 | 60 | |
XM_017021817.1 | XP_016877306.1 | 1 - 2 | 74 | 60 | |
GenBank mRNA | DB017355.1 | - | 1 - 2 | 74 | 60 |