The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130852.1 | NP_001124324.1 | 8 - 9 | 1284 | 108 |
NM_012116.3 | NP_036248.3 | 9 - 10 | 1422 | 108 | |
GenBank mRNA | AB028645.1 | - | 9 - 10 | 1400 | 108 |
AF117646.1 | - | 9 - 10 | 1422 | 108 | |
AF117647.1 | - | 8 - 9 | 1284 | 108 | |
BC028915.1 | - | 9 - 10 | 1380 | 108 |