Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC3
Entrez Gene ID
219681
Gene Name
armadillo repeat containing 3
Gene Aliases
CT81, KU-CT-1
Chromosome Location
Chr.10: 22928024 - 23038527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282745.1 NP_001269674.1 7 - 8 810 153
NM_001282746.1 NP_001269675.1 7 - 8 810 153
NM_173081.4 NP_775104.2 7 - 8 810 153
XM_005252381.2 XP_005252438.1 6 - 7 631 153
XM_011519350.2 XP_011517652.1 7 - 8 793 153
XM_017015832.1 XP_016871321.1 7 - 8 793 153
GenBank mRNA AK057389.1 - 7 - 8 753 153
AK302985.1 - 7 - 8 797 153
AY436643.1 - 7 - 8 771 153
BC039312.1 - 7 - 8 803 153

View Full Product Details