Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CMAS
Entrez Gene ID
55907
Gene Name
cytidine monophosphate N-acetylneuraminic acid synthetase
Gene Aliases
CSS
Chromosome Location
Chr.12: 22046174 - 22065674 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018686.5 NP_061156.1 2 - 3 526 139
NR_135117.1 2 - 3 526 139
GenBank mRNA AF271388.1 - 2 - 3 414 139
AK022927.1 - 2 - 3 436 139
AK222981.1 - 2 - 3 425 139
AL832975.1 - 2 - 3 474 139
BC016609.1 - 2 - 3 475 139

View Full Product Details