The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032565.4 | NP_115954.1 | 3 - 4 | 446 | 81 |
NR_103802.1 | 4 - 5 | 567 | 81 | ||
NR_103803.1 | 2 - 3 | 376 | 81 | ||
GenBank mRNA | AF243433.1 | - | 3 - 4 | 435 | 81 |
BC018478.1 | - | 3 - 4 | 412 | 81 | |
BC073152.1 | - | 4 - 5 | 567 | 81 | |
BC092471.1 | - | 3 - 4 | 441 | 81 | |
BY799168.2 | - | 2 - 3 | 342 | 81 |