Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RPL13AP17
- Entrez Gene ID
-
399670
- Gene Name
- ribosomal protein L13a pseudogene 17
- Gene Aliases
-
RPL13A_6_816
- Chromosome Location
-
Chr.7: 78347242 - 78359458 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003680.1 |
|
1 - 2 |
83 |
149 |
GenBank mRNA |
BC027852.1 |
- |
1 - 2 |
83 |
149 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB003300.1 |
- |
1 - 2 |
184 |
|
Chr. 7 - 78347142 - 78351898 [+] on Build GRCh38 |
149 |
GenBankmRNA |
DB001703.1 |
- |
1 - 2 |
89 |
|
Chr. 7 - 78347236 - 78350529 [+] on Build GRCh38 |
149 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105375368 |
LOC105375368 |
|
-350.9kb
|
Sense |
Chr. - 77990374 - 77996366 on Build GRCh38 |
Downstream |
MAGI2 antisense RNA 2 |
MAGI2-AS2 |
|
660.6kb
|
Sense |
Chr. - 79008988 - 79012277 on Build GRCh38 |
View Full Product Details