The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000197.1 | NP_000188.1 | 9 - 10 | 721 | 125 |
XM_011518618.2 | XP_011516920.1 | 12 - 13 | 1193 | 125 | |
XM_011518619.2 | XP_011516921.1 | 11 - 12 | 1031 | 125 | |
XM_017014671.1 | XP_016870160.1 | 11 - 12 | 3105 | 125 | |
XM_017014672.1 | XP_016870161.1 | 11 - 12 | 3029 | 125 | |
XM_017014673.1 | XP_016870162.1 | 10 - 11 | 3070 | 125 | |
XM_017014674.1 | XP_016870163.1 | 10 - 11 | 3000 | 125 | |
XM_017014675.1 | XP_016870164.1 | 10 - 11 | 2786 | 125 | |
XM_017014676.1 | XP_016870165.1 | 11 - 12 | 3145 | 163 | |
XM_017014677.1 | XP_016870166.1 | 12 - 13 | 3686 | 125 | |
GenBank mRNA | BC034281.1 | - | 9 - 10 | 705 | 125 |
CR541894.1 | - | 9 - 10 | 673 | 125 | |
KU178130.1 | - | 9 - 10 | 673 | 125 | |
KU178131.1 | - | 10 - 11 | 851 | 125 | |
KU178132.1 | - | 9 - 10 | 673 | 125 | |
KU178133.1 | - | 7 - 8 | 602 | 125 | |
KU178134.1 | - | 4 - 5 | 350 | 125 | |
U05659.1 | - | 9 - 10 | 721 | 125 |